Alfa Aesar™ Lead(II) oxalate, Puratronic™, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 1299906

Quantity 5g

 View more versions of this product

Catalog No. AA1299906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 814-93-7
    Linear Formula PbC2O4
    Formula Weight 295.23
    MDL Number MFCD00040677
    UN Number UN2291
    Chemical Name or Material Lead(II) oxalate
    Grade Puratronic™

    Specifications

    Formula Weight 295.23
    Health Hazard 3 GHS P Statement
    P273-P201-P308+P313-P501a
    Avoid release to the environment.
    Obtain special instructions before use.
    IF exposed or concerned: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    MDL Number MFCD00040677
    CAS 814-93-7
    Melting Point 300°C (dec.)
    TSCA TSCA
    Physical Form Powder
    UN Number UN2291
    Quantity 5g
    Health Hazard 1 Danger
    Linear Formula PbC2O4
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.
    Health Hazard 2 GHS H Statement
    H360-H373-H302-H332
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.
    Chemical Name or Material Lead(II) oxalate
    Assay 99.999%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H360-H373-H302-H332
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P273-P201-P308+P313-P501a
    Avoid release to the environment.
    Obtain special instructions before use.
    IF exposed or concerned: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit