Lead(II) oxide, 99.99% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1424030

Quantity 250g

 View more versions of this product

Catalog No. AA1424030

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1317-36-8
    Physical Form Powder
    Assay Percent Range 99.99% (Metal basis)
    Molecular Formula OPb
    Formula Weight 223.19
    MDL Number MFCD00011164
    Chemical Name or Material Lead(II) oxide

    Specifications

    Physical Form Powder
    Formula Weight 223.19
    Quantity 250g
    Color Yellow
    DOT Information DOT Class: 9, Packaging Group: III
    Density 9.53g/cm3 at 20°C
    EINECS Number 215-267-0
    Odor Odorless
    Assay Percent Range 99.99% (Metal basis)
    Molecular Formula OPb
    Appearance Yellow powder
    MDL Number MFCD00011164
    CAS 1317-36-8
    Health Hazard 1 H360-H373-H400-H410-H302-H332: May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful if inhaled.
    Chemical Name or Material Lead(II) oxide

    H360-H373-H400-H410-H302-H332: May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful if inhaled.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit