Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Lead(II) oxide, low silver
Click to view available options
Quantity:
500g
2kg
Description
- As storage batteries, ceramic cements and fluxes, pottery and glazes, in chromium pigments, in oil refining, in paints, and in the assay of precious metal ores
Specifications
UN Number | UN3077 |
Merck Index | 14,5413 |
Solubility Information | Insoluble in water and alcohol; Soluble in acetic acid,dilute HNO3 and alkalies |
Formula Weight | 223.19 |
Physical Form | Powder |
Density | 9.53 g/mL |
Odor | Odorless |
Melting Point | 886°C |
Quantity | 2kg |
Chemical Name or Material | Lead(II) oxide, Low silver |
Safety and Handling
GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
H302+H332-H350-H360Df-H372
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 215-267-0
RTECSNumber : OG1750000
TSCA : Yes
Recommended Storage : Ambient temperatures