Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Fluka™ Lead(II) oxide, meets analytical spec. of DAC, BPC, 99 to 100.5% (complexometric), yellow, Honeywell Fluka™

Click to view available options
Quantity:
1kg
25kg
Packaging:
Fiberboard Box
Poly bottle
1317-36-8
OPb
223.199
MFCD00011164
YEXPOXQUZXUXJW-UHFFFAOYSA-N
lead monoxide,lead ii oxide,lead oxide pbo,massicot,massicotite,lead monooxide,lead protoxide,plumbous oxide,litharge pure,lead oxide yellow
14827
oxolead
O=[Pb]
1317-36-8
OPb
223.199
MFCD00011164
YEXPOXQUZXUXJW-UHFFFAOYSA-N
lead monoxide,lead ii oxide,lead oxide pbo,massicot,massicotite,lead monooxide,lead protoxide,plumbous oxide,litharge pure,lead oxide yellow
14827
oxolead
O=[Pb]

Meets analytical specification of DAC, BPC, 99-100.5% (complexometric), yellow

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Linear Formula PbO
UN Number 2291
Formula Weight 223.2g/mol
Percent Purity 99 to 100.5% (complexometric)
Packaging Poly bottle
Vapor Pressure 10mmHg (0°C)
Color Yellow
Quantity 1kg
Chemical Name or Material Lead(Ii) Oxide, Meets analytical specification of DAC, BPC

P201-P260-P263-P280-P301 + P312 + P330-P308 + P313

H302 + H332-H351-H360Df-H362-H372-H410

EINECSNumber : 215-267-0

RTECSNumber : OG1750000