Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(II) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog Number AA4075322
Click to view available options
Quantity:
25 g
100 g
500 g
1317-36-8
OPb
223.20
MFCD00011164
YEXPOXQUZXUXJW-UHFFFAOYSA-N
lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow
14827
oxolead
O=[Pb]
1317-36-8
OPb
223.20
MFCD00011164
YEXPOXQUZXUXJW-UHFFFAOYSA-N
lead monoxide, lead ii oxide, lead oxide pbo, massicot, massicotite, lead monooxide, lead protoxide, plumbous oxide, litharge pure, lead oxide yellow
14827
oxolead
O=[Pb]

Used in producing iridescent colors on brass and bronze, coloring sulfur-containing substances and used as pigment for rubber. It is also used in oil refining, varnishes, paints, enamels and in assays for precious metal ores.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 886°C
Color Yellow
Physical Form Powder
Quantity 100 g
UN Number UN3077
Merck Index 14,5413
Solubility Information Insoluble in water,soluble in concentrated alkalies,hydrochloric acid and ammonium chloride.
Formula Weight 223.19
Percent Purity 99.999%
Grade Puratronic™
Odor Odorless
Density 9.53 g/mL
Assay (metals basis)
Chemical Name or Material Lead(II) oxide
Show More Show Less
Hazard Category H302+H332-H350-H360Df-H372
Hazard Statement GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 215-267-0
RTECSNumber OG1750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit