Learn More
Lead(II) oxide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Description
Used in producing iridescent colors on brass and bronze, coloring sulfur-containing substances and used as pigment for rubber. It is also used in oil refining, varnishes, paints, enamels and in assays for precious metal ores.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 886°C |
Color | Yellow |
Physical Form | Powder |
Quantity | 500 g |
UN Number | UN3077 |
Merck Index | 14,5413 |
Solubility Information | Insoluble in water,soluble in concentrated alkalies,hydrochloric acid and ammonium chloride. |
Formula Weight | 223.19 |
Percent Purity | 99.999% |
Grade | Puratronic™ |
Show More |
Safety and Handling
Hazard Category | H302+H332-H350-H360Df-H372 |
Hazard Statement | GHS H Statement H360-H373-H302-H332 May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Harmful if swallowed. Harmful if inhaled. |
Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c |
DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
EINECSNumber | 215-267-0 |
RTECSNumber | OG1750000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.