Alfa Aesar™ Lead(II) oxide, Puratronic™, 99.9995% (metals basis)

Manufacturer: Alfa Aesar™ 1072836

Quantity 500g

 View more versions of this product

Catalog No. AA1072836

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1317-36-8
    Linear Formula PbO
    Formula Weight 223.19
    MDL Number MFCD00011164
    UN Number UN3077
    Merck Index 14,5413
    Chemical Name or Material Lead(II) oxide
    Grade Puratronic™

    Specifications

    Formula Weight 223.19
    Health Hazard 3 GHS P Statement
    P260-P261-P281-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Merck Index 14,5413
    Health Hazard 2 GHS H Statement
    H360-H373-H302-H332
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.
    TSCA TSCA
    Odor Odorless
    Physical Form Powder
    MDL Number MFCD00011164
    UN Number UN3077
    Melting Point 886°C
    Quantity 500g
    Health Hazard 1 Danger
    Density 9.53g/mL
    CAS 1317-36-8
    Color Yellow
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    RTECS Number OG1750000
    Linear Formula PbO
    Chemical Name or Material Lead(II) oxide
    Assay 99.9995%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H360-H373-H302-H332
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P260-P261-P281-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit