Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(II) perchlorate trihydrate, ACS reagent, Acros Organics

Click to view available options
Quantity:
100g
500g
Packaging:
Plastic bottle
13453-62-8
Cl2O8Pb·3H2O
460.14
KKGGEAQRICYXNM-UHFFFAOYSA-L
lead ii perchlorate trihydrate,acmc-20aknd,2clo4.pb.3h2o,lead perchlorate 3h2o,lead ii perchlorate, trihydrate,lead perchlorate, trihydrate, reagent acs,lead ii perchlorate trihydrate, acs reagent
25021834
lead(2+);diperchlorate;trihydrate
O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Pb+2]
13453-62-8
Cl2O8Pb·3H2O
460.14
KKGGEAQRICYXNM-UHFFFAOYSA-L
lead ii perchlorate trihydrate,acmc-20aknd,2clo4.pb.3h2o,lead perchlorate 3h2o,lead ii perchlorate, trihydrate,lead perchlorate, trihydrate, reagent acs,lead ii perchlorate trihydrate, acs reagent
25021834
lead(2+);diperchlorate;trihydrate
O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Pb+2]
Linear Formula Pb(ClO4)2·3H2O
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
Formula Weight 460.14
Grade ACS Reagent
Chloride 0.01% max
Density 2.6000g/mL
Insoluble Matter 0.005% max
Packaging Plastic bottle
Solubility Solubility in water: soluble. Other solubilities: soluble in alcohol
Specific Gravity 2.6
Melting Point 100.0°C
Quantity 100g
Chemical Name or Material Lead(II) perchlorate trihydrate
Show More Show Less

GHS Signal Word: Danger