Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Lead(II) selenate, 99.9%

Manufacturer:  Alfa Aesar™ 3934514

Quantity 25g

 View more versions of this product

Catalog No. AA3934514

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

7446-15-3
MFCD00054013
Lead(II) selenate
350.16
UN2630

Specifications

7446-15-3
350.16
UN2630
6.37g/mL
Lead(II) selenate
Powder
MFCD00054013
Insoluble in water; Soluble in concentrated acids
25g
99.9%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H300-H330-H360-H373
Fatal if swallowed.
Fatal if inhaled.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: SELENATES

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit