Alfa Aesar™ Lead(II) selenate, 99.9%

Manufacturer: Alfa Aesar™ 3934506

Quantity 5g

 View more versions of this product

Catalog No. AA3934506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7446-15-3
    Formula Weight 350.16
    MDL Number MFCD00054013
    UN Number UN2630
    Chemical Name or Material Lead(II) selenate

    Specifications

    Formula Weight 350.16
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H300-H330-H360-H373
    Fatal if swallowed.
    Fatal if inhaled.
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Physical Form Powder
    MDL Number MFCD00054013
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: SELENATES
    UN Number UN2630
    Solubility Information Insoluble in water; Soluble in concentrated acids
    Density 6.37g/mL
    TSCA TSCA
    CAS 7446-15-3
    Quantity 5g
    Chemical Name or Material Lead(II) selenate
    Assay 99.9%

    GHS H Statement
    H300-H330-H360-H373
    Fatal if swallowed.
    Fatal if inhaled.
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit