Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lead(II) selenide, 99%, Thermo Scientific™
Click to view available options
Quantity:
10 g
50 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Melting Point | 1,065°C |
Physical Form | Pieces |
Quantity | 10 g |
UN Number | UN3077 |
Formula Weight | 286.15 |
Percent Purity | 99% |
Odor | Product specific |
Chemical Name or Material | Lead(II) selenide |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | GHS H Statement H300-H330-H360-H373 Fatal if swallowed. Fatal if inhaled. May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. |
Precautionary Statement | GHS P Statement P260-P301+P310-P304+P340-P320-P330-P405-P501a Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Specific treatment is urgent (see label). Rinse mouth. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
TSCA | TSCA |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction