Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(II) sulfate, 99%, Acros Organics

Click to view available options
Quantity:
100g
500g
Packaging:
Plastic bottle
7446-14-2
O4PbS
303.25
MFCD00011166
KEQXNNJHMWSZHK-UHFFFAOYSA-L
lead ii sulfate,lead sulfate,anglislite,lead sulphate,mulhouse white,lead bottoms,lead monosulfate,fast white,milk white,natural anglesite
24008
lead(2+);sulfate
[O-]S(=O)(=O)[O-].[Pb+2]
7446-14-2
O4PbS
303.25
MFCD00011166
KEQXNNJHMWSZHK-UHFFFAOYSA-L
lead ii sulfate,lead sulfate,anglislite,lead sulphate,mulhouse white,lead bottoms,lead monosulfate,fast white,milk white,natural anglesite
24008
lead(2+);sulfate
[O-]S(=O)(=O)[O-].[Pb+2]
Assay Percent Range 99%
Merck Index 15, 5475
Solubility Information Solubility in water: slightly soluble
Formula Weight 303.25
Physical Form Crystalline Powder
Percent Purity 99%
Elemental Analysis 98.5% min. (AES)
Identification Confirmed
Packaging Plastic bottle
Solubility Solubility in water: slightly soluble.
Color White
Melting Point 1170.0°C
Quantity 500g
Chemical Name or Material Lead(II) sulfate
Show More Show Less

GHS H Statement
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
May damage the unborn child.
Suspected

GHS P Statement
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a pos

GHS Signal Word: Danger

EINECSNumber : 231-198-9