Alfa Aesar™ Lead(II) sulfate, Puratronic™, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 1073009

Quantity 10g

 View more versions of this product

Catalog No. AA1073009

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7446-14-2
    Formula Weight 303.25
    MDL Number MFCD00011166
    UN Number UN2291
    Merck Index 14,5420
    Chemical Name or Material Lead(II) sulfate
    Grade Puratronic™

    Specifications

    Formula Weight 303.25
    Health Hazard 3 GHS P Statement
    P260-P261-P281-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00011166
    UN Number UN2291
    Quantity 10g
    TSCA TSCA
    Odor Odorless
    Health Hazard 1 Danger
    Physical Form Crystalline
    Merck Index 14,5420
    Density 6.2g/mL
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.
    CAS 7446-14-2
    Melting Point 1170°C
    Health Hazard 2 GHS H Statement
    H360-H373-H302-H332
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.
    Chemical Name or Material Lead(II) sulfate
    Assay 99.999%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H360-H373-H302-H332
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P260-P261-P281-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit