Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead(II) sulfate, Reagent Grade, Thermo Scientific Chemicals

Catalog Number p-4407549
Click to view available options
Quantity:
100 g
500 g
2.5 kg
7446-14-2
O4PbS
303.256
MFCD00011166
KEQXNNJHMWSZHK-UHFFFAOYSA-L
lead ii sulfate, lead sulfate, anglislite, lead sulphate, mulhouse white, lead bottoms, lead monosulfate, fast white, milk white, natural anglesite
24008
lead(2+);sulfate
[O-]S(=O)(=O)[O-].[Pb+2]
7446-14-2
O4PbS
303.256
MFCD00011166
KEQXNNJHMWSZHK-UHFFFAOYSA-L
lead ii sulfate, lead sulfate, anglislite, lead sulphate, mulhouse white, lead bottoms, lead monosulfate, fast white, milk white, natural anglesite
24008
lead(2+);sulfate
[O-]S(=O)(=O)[O-].[Pb+2]

For Use in Pigments, Galvanic Batteries, Lithography

Lead(II) sulfate is used in pigments, galvanic batteries, lithography, weighting fabrics. It is also used as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 1170°C
Color White
Physical Form Lump/Powder
Quantity 100 g
Solubility Information Partially soluble in water,dilute HCl and HNO3. Soluble in sodium hydroxide,ammonium acetate or tartrate,concentrated hydriodic acid. Insoluble in alcohol
Formula Weight 303.25
Grade Reagent
Odor Odorless
Appearance White powder/lump
Density 6.2 g/cm3 at 20°C
Chemical Name or Material Lead(II) sulfate
Show More Show Less
Hazard Category H302+H312+H332-H350-H360-H373
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
DOTInformation DOT Class: 9; Packaging Group: III
EINECSNumber 231-198-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit