Lead(II) sulfide, 99.995% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 4546204

Quantity 2g

 View more versions of this product

Catalog No. AA4546204

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1314-87-0
    Physical Form Lumps
    Assay Percent Range 99.995% (Metal basis)
    Molecular Formula PbS
    Formula Weight 239.25
    MDL Number MFCD00016280
    Chemical Name or Material Lead(II) sulfide

    Specifications

    Physical Form Lumps
    Formula Weight 239.25
    Quantity 2g
    Color Black
    DOT Information DOT Class: 9, Packaging Group: III
    Density 7.5g/cm3 at 20°C
    EINECS Number 215-246-6
    Odor Odorless
    Assay Percent Range 99.995% (Metal basis)
    Molecular Formula PbS
    Appearance Black lumps
    MDL Number MFCD00016280
    CAS 1314-87-0
    Health Hazard 1 H360-H373-H400-H410-H302-H332: May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful if inhaled.
    Chemical Name or Material Lead(II) sulfide

    H360-H373-H400-H410-H302-H332: May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful if inhaled.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit