Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Lead(II) tantalum oxide, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 3934814

Quantity 25g

 View more versions of this product

Catalog No. AA3934814


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

12065-68-8
MFCD00054256
Lead(II) tantalate
665.09
UN2291
Lead(II) tantalum oxide

Specifications

12065-68-8
665.09
UN2291
7.9g/mL
>350°C
Lead(II) tantalum oxide
Powder
MFCD00054256
Lead(II) tantalate
−200 Mesh
25g
99.9%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit