Alfa Aesar™ Lead(II) tartrate, 99%

Manufacturer: Alfa Aesar™ A1238814

Quantity 25g

 View more versions of this product

Catalog No. AAA1238814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 815-84-9
    Molecular Formula C4H4O6Pb
    Formula Weight 355.27
    MDL Number MFCD00043282
    UN Number UN2291
    Synonym Tartaric acid lead(II) salt
    Chemical Name or Material Lead (II) tartrate

    Specifications

    Formula Weight 355.27
    Health Hazard 3 GHS P Statement
    P260-P261-P281-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 815-84-9
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H360-H373-H302-H332
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.
    TSCA TSCA
    Molecular Formula C4H4O6Pb
    MDL Number MFCD00043282
    UN Number UN2291
    Synonym Tartaric acid lead(II) salt
    Quantity 25g
    RTECS Number OG4800000
    Chemical Name or Material Lead (II) tartrate
    Assay 99%

    Description

    Applications
    Used as a raw material in organic chemistry for the synthesis of other chiral molecules. The acid has served in the farming and metal industries as a chelating agent for complexing micronutrients.

    Solubility
    Soluble in dilute nitric acid, alkali, acid ammonium hydrogen oxidation and tartaric acid ammonium solution, insoluble in water and alcohol.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    GHS H Statement
    H360-H373-H302-H332
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P260-P261-P281-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit