Alfa Aesar™ Lead(II) thiocyanate, 99.99% (metals basis)

Manufacturer: Alfa Aesar™ 44084-14

Quantity 25g

 View more versions of this product

Catalog No. AA4408414

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 592-87-0
    Molecular Formula Pb(SCN)2
    Formula Weight 323.35
    MDL Number MFCD00011155
    UN Number UN2291
    Beilstein 3687984
    Merck Index 14,5426
    Chemical Name or Material Lead (II) thiocyanate

    Specifications

    Formula Weight 323.35
    Odor Odorless
    CAS 592-87-0
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H331-H360-H373-H302-H312
    Toxic if inhaled.
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful in contact with skin.
    Health Hazard 3 GHS P Statement
    P260-P261-P280-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN
    Molecular Formula Pb(SCN)2
    MDL Number MFCD00011155
    Quantity 25g
    Density 3.82g/mL
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.
    TSCA TSCA
    Physical Form Powder
    UN Number UN2291
    Beilstein 3687984
    Merck Index 14,5426
    Melting Point 190°C
    Chemical Name or Material Lead (II) thiocyanate
    Assay 99.99%, (metals basis)

    GHS H Statement
    H331-H360-H373-H302-H312
    Toxic if inhaled.
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P260-P261-P280-P304+P340-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit