Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Lead(II) thiocyanate, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 4408422

Quantity 100g

 View more versions of this product

Catalog No. AA4408422

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

592-87-0
323.35
UN2291
14,5426
Pb(SCN)2
MFCD00011155
3687984
Lead (II) thiocyanate

Specifications

592-87-0
Pb(SCN)2
MFCD00011155
3687984
3.82g/mL
190°C
Lead (II) thiocyanate
Powder
323.35
UN2291
14,5426
Odorless
100g
99.99%, (metals basis)

Safety and Handling

GHS H Statement
H331-H360-H373-H302-H312

GHS P Statement
P260-P201-P261-P280-P281-P304+P340-P311-P301+P312-P405-P501a

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit