Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Lead(II) titanium(IV) 2-ethylhexanoate tetraisopropoxide, 99% (metals basis), 50% w/v in isopropanol

Manufacturer:  Alfa Aesar™ 36576AC

Quantity 25mL

 View more versions of this product

Catalog No. AA36576AC


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

777.4
MFCD00171336
UN1993
Lead(II) titanium(IV) 2-ethylhexanoate tetraisopropoxide

Specifications

25mL
Moisture Sensitive
777.4
UN1993
99%, (metals basis)
50% w/v in isopropanol
Liquid
MFCD00171336
Lead(II) titanium(IV) 2-ethylhexanoate tetraisopropoxide

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H360-H373-H302-H319-H336
Highly flammable liquid and vapor.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Causes serious eye irritation.
May cause drowsiness or dizziness.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit