Alfa Aesar™ Lead(II) titanium oxide, 99.5%

Manufacturer: Alfa Aesar™ 14139A3

Quantity 2kg

 View more versions of this product

Catalog No. AA14139A3

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12060-00-3
    Formula Weight 303.09
    MDL Number MFCD00011159
    UN Number UN3288
    Synonym Lead(II) titanate
    Chemical Name or Material Lead(II) titanium oxide

    Specifications

    Formula Weight 303.09
    Health Hazard 3 GHS P Statement
    P260-P261-P281-P301+P310-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    MDL Number MFCD00011159
    Color Yellow
    Quantity 2kg
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H300-H360-H373-H332
    Fatal if swallowed.
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if inhaled.
    TSCA TSCA
    Mesh Size −325 Mesh
    Physical Form Powder
    CAS 12060-00-3
    UN Number UN3288
    Density 7.520g/mL
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
    Synonym Lead(II) titanate
    RTECS Number XR1575000
    Chemical Name or Material Lead(II) titanium oxide
    Assay 99.5%

    GHS H Statement
    H300-H360-H373-H332
    Fatal if swallowed.
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if inhaled.

    GHS P Statement
    P260-P261-P281-P301+P310-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Use personal protective equipment as required.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit