Alfa Aesar™ Lead(II) trifluoroacetate hemihydrate

Manufacturer: Alfa Aesar™ 1755318

Quantity 50g

 View more versions of this product

Catalog No. AA1755318

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4146-73-0
    Linear Formula Pb(CF3COO)2·0.5H2O
    Formula Weight 442.23 (433.22 Anhydrous)
    MDL Number MFCD00061594
    UN Number UN2291
    Chemical Name or Material Lead(II) trifluoroacetate hemihydrate

    Specifications

    Formula Weight 442.23 (433.22 Anhydrous)
    Health Hazard 3 GHS P Statement
    P273-P201-P308+P313-P501a
    Avoid release to the environment.
    Obtain special instructions before use.
    IF exposed or concerned: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 147° to 153°C
    Quantity 50g
    MDL Number MFCD00061594
    Odor Odorless
    Health Hazard 2 GHS H Statement
    H360-H373-H302-H332
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.
    CAS 4146-73-0
    UN Number UN2291
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.
    Physical Form Powder
    Linear Formula Pb(CF3COO)2·0.5H2O
    Color White
    Chemical Name or Material Lead(II) trifluoroacetate hemihydrate

    GHS H Statement
    H360-H373-H302-H332
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P273-P201-P308+P313-P501a
    Avoid release to the environment.
    Obtain special instructions before use.
    IF exposed or concerned: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit