Lead(IV) oxide, Brown to black powder, 97.0% min., Alfa Aesar™

For Use in Battery Electrodes, Manufacturing of Pigments and Dyes, As Analytical Lab Reagent

Manufacturer: Alfa Aesar 12483A3

Quantity 2kg

 View more versions of this product

Catalog No. AA12483A3

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1309-60-0
    Physical Form Powder
    Assay Percent Range 97.0% min.
    Molecular Formula PbO2
    Formula Weight 239.19
    MDL Number MFCD00011165
    Synonym Lead dioxide
    Chemical Name or Material Lead(IV) oxide
    Grade ACS Grade

    Specifications

    Physical Form Powder
    Formula Weight 239.19
    Quantity 2kg
    Color Brown to black
    DOT Information DOT Class: 5.1, Packaging Group: III
    EINECS Number 215-174-5
    Odor Odorless
    Assay Percent Range 97.0% min.
    Molecular Formula PbO2
    MDL Number MFCD00011165
    CAS 1309-60-0
    Synonym Lead dioxide
    Health Hazard 1 H360-H373-H272-H400-H410-H302-H332: May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. May intensify fire; oxidizer. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful if inhaled.
    Chemical Name or Material Lead(IV) oxide
    Grade ACS Grade

    H360-H373-H272-H400-H410-H302-H332: May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. May intensify fire; oxidizer. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful if inhaled.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit