Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Lead lanthanum zirconium titanium oxide, polymeric precursor

Piezoelectric and electro-optic devices

Manufacturer:  Alfa Aesar™ 4473403

Quantity 1g

 View more versions of this product

Catalog No. AA4473403


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

MFCD02684260
UN1993
Lead lanthanum zirconium titanium oxide, Polymeric precursor

Specifications

Viscous Liquid
UN1993
Moisture Sensitive
1g
MFCD02684260
Soluble in hexane, toluene and alcohol
Brown
Lead lanthanum zirconium titanium oxide, Polymeric precursor

Safety and Handling

SDS & Safety Documents

GHS H Statement
H360-H373-H226-H302-H332-H336
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P273-P201-P308+P313-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit