Alfa Aesar™ Lead lanthanum zirconium titanium oxide, polymeric precursor

Piezoelectric and electro-optic devices

Manufacturer: Alfa Aesar™ 4473414

Quantity 25g

 View more versions of this product

Catalog No. AA4473414

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD02684260
    UN Number UN1993
    Chemical Name or Material Lead lanthanum zirconium titanium oxide, Polymeric precursor

    Specifications

    Physical Form Viscous Liquid
    Health Hazard 3 GHS P Statement
    P273-P201-P308+P313-P501a
    Avoid release to the environment.
    Obtain special instructions before use.
    IF exposed or concerned: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Solubility Information Soluble in hexane, toluene and alcohol
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Sensitivity Moisture Sensitive
    MDL Number MFCD02684260
    UN Number UN1993
    Health Hazard 2 GHS H Statement
    H360-H373-H226-H302-H332-H336
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful if inhaled.
    Color Brown
    Quantity 25g
    Chemical Name or Material Lead lanthanum zirconium titanium oxide, Polymeric precursor

    GHS H Statement
    H360-H373-H226-H302-H332-H336
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P273-P201-P308+P313-P501a
    Avoid release to the environment.
    Obtain special instructions before use.
    IF exposed or concerned: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit