Learn More
Research Products International CorpSupplier Diversity Partner Lead Waste Container with Lift Off Lid, Small DFS Item
Manufacturer:  Research Products International CorpSupplier Diversity Partner 050200
Lead lined waste containers provide safe storage for items contaminated with Iodine-125. Featuring a stainless steel exterior with a 1/8 inch thick lead-lined interior. Benchtop size container is convenient for disposal of items such as syringes, vials, ampoules, etc. Diameter: 6.5 x 5 Inches. Lead Shielding: .125 Inches Thick
Catalog No. 50-136-7620