Alfa Aesar™ Lead molybdenum oxide, 99%

Manufacturer: Alfa Aesar™ A1057622

Quantity 100g

 View more versions of this product

Catalog No. AAA1057622

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10190-55-3
    Molecular Formula MoO4Pb
    Linear Formula PbMoO4
    Formula Weight 367.13
    MDL Number MFCD00016272
    UN Number UN3077
    Synonym Lead molbydate
    Chemical Name or Material Lead molybdenum oxide

    Specifications

    Formula Weight 367.13
    Health Hazard 3 GHS P Statement
    P273-P201-P308+P313-P501a
    Avoid release to the environment.
    Obtain special instructions before use.
    IF exposed or concerned: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    MDL Number MFCD00016272
    Odor Odorless
    Molecular Formula MoO4Pb
    UN Number UN3077
    Synonym Lead molbydate
    Quantity 100g
    Health Hazard 2 GHS H Statement
    H360-H373-H302-H332
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.
    Linear Formula PbMoO4
    CAS 10190-55-3
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Chemical Name or Material Lead molybdenum oxide
    Assay 99%

    GHS H Statement
    H360-H373-H302-H332
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P273-P201-P308+P313-P501a
    Avoid release to the environment.
    Obtain special instructions before use.
    IF exposed or concerned: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit