Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Lead naphthenate, 61% min in mineral spirits, Pb 20%

Manufacturer:  Alfa Aesar™ 38593A1

Quantity 1kg

 View more versions of this product

Catalog No. AA38593A1


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

61790-14-5
MFCD00466946
UN1993
Lead naphthenate

Specifications

1kg
61% min. in min.eral spirits, Pb 20%
Brown
Liquid
UN1993
61790-14-5
1.13g/mL
40° (104°F)
MFCD00466946
Lead naphthenate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H360-H373-H226-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P210-P260-P261-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit