Alfa Aesar™ Lead naphthenate, 61% min in mineral spirits, Pb 20%

Manufacturer: Alfa Aesar™ 3859330

Quantity 250g

 View more versions of this product

Catalog No. AA3859330

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 61790-14-5
    MDL Number MFCD00466946
    UN Number UN1993
    Chemical Name or Material Lead naphthenate

    Specifications

    Health Hazard 3 GHS P Statement
    P210-P260-P261-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Quantity 250g
    Health Hazard 1 Danger
    Density 1.13g/mL
    Health Hazard 2 GHS H Statement
    H360-H373-H226-H302-H332
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful if inhaled.
    Flash Point 40° (104°F)
    MDL Number MFCD00466946
    CAS 61790-14-5
    UN Number UN1993
    Concentration 61% min. in min.eral spirits, Pb 20%
    Color Brown
    TSCA TSCA
    Physical Form Liquid
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Chemical Name or Material Lead naphthenate

    GHS H Statement
    H360-H373-H226-H302-H332
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P210-P260-P261-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit