Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead Nitrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle
10099-74-8
N2O6Pb
331.208
RLJMLMKIBZAXJO-UHFFFAOYSA-N
lead dinitrate,lead nitrate,lead ii nitrate,plumbous nitrate,lead 2+ nitrate,lead nitrate pb no3 2,lead ii nitrate,nitrate de plomb french,lead ii nitrate 1:2,nitric acid, lead 2+ salt
24924
CHEBI:37187
lead(2+);dinitrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Pb+2]
10099-74-8
N2O6Pb
331.208
RLJMLMKIBZAXJO-UHFFFAOYSA-N
lead dinitrate,lead nitrate,lead ii nitrate,plumbous nitrate,lead 2+ nitrate,lead nitrate pb no3 2,lead ii nitrate,nitrate de plomb french,lead ii nitrate 1:2,nitric acid, lead 2+ salt
24924
CHEBI:37187
lead(2+);dinitrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Pb+2]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 331.2
Physical Form Crystals
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 4.53kg
Quantity 500g
Chemical Name or Material Lead Nitrate