Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead Nitrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle

Chemical Identifiers

10099-74-8
N2O6Pb
331.208
RLJMLMKIBZAXJO-UHFFFAOYSA-N
24924
CHEBI:37187

High quality chemicals for laboratory and specialized industrial use

SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Pb+2]
IUPAC Name lead(2+);dinitrate
Formula Weight 331.2
Physical Form Crystals
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 4.53kg
Quantity 125g
Chemical Name or Material Lead Nitrate
Grade Baker Analyzed™ ACS Reagent