Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead Oxide, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
2.5kg
500g
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle

Chemical Identifiers

1317-36-8
OPb
223.199
YEXPOXQUZXUXJW-UHFFFAOYSA-N
Litharge
14827

High quality chemicals for laboratory and specialized industrial use

CAS 1317-36-8
Molecular Formula OPb
Synonym Litharge
InChI Key YEXPOXQUZXUXJW-UHFFFAOYSA-N
SMILES O=[Pb]
IUPAC Name oxolead
Molecular Weight (g/mol) 223.199
PubChem CID 14827
Formula Weight 223.19
Packaging Poly Bottle
Specific Gravity 1L = 9.53kg
Quantity 500g
Chemical Name or Material Lead Oxide
Grade BAKER ANALYZED™ Reagent
Show More Show Less