Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lead powder, -100 mesh, 99.95% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Black to Gray |
Appearance | Gray to black atomized powder |
Density | 11.34g/cm3 at 20°C |
Assay Percent Range | 99.95% (Metals basis) |
Quantity | 250 g |
Mesh Size | -100 Mesh |
Odor | Odorless |
Form | Atomized Powder |
Chemical Name or Material | Lead |
Safety and Handling
Hazard Category | H350-H360FD-H362-H372 |
Hazard Statement | Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets. |
Precautionary Statement | P201-P202-P260-P263-P264b-P270-P281-P308+P313-P501c |
DOTInformation | DOT Class: 9, Packaging Group: III |
EINECSNumber | 231-100-4 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction