Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lead powder, 99% (metals basis), Alfa Aesar™

Click to view available options
Quantity:
100g
500g
2kg
7439-92-1
Pb
207.2
MFCD00134050
WABPQHHGFIMREM-UHFFFAOYSA-N
plumbum,metal,blei,lead, elemental,rough bullion,plomo,glover,element,flake,omaha & grant
5352425
CHEBI:27889
lead
[Pb]
7439-92-1
Pb
207.2
MFCD00134050
WABPQHHGFIMREM-UHFFFAOYSA-N
plumbum,metal,blei,lead, elemental,rough bullion,plomo,glover,element,flake,omaha & grant
5352425
CHEBI:27889
lead
[Pb]
Quantity 2kg
Appearance Gray to black powder
Assay Percent Range 99% (Metals basis)
Mesh Size -200 Mesh
Odor Odorless
Physical Form Powder
Color Black to Gray
Chemical Name or Material Lead

P201-P202-P260-P263-P264b-P270-P281-P308+P313-P501c

H350-H360FD-H362-H372

DOTInformation : DOT Class: 9, Packaging Group: III

EINECSNumber : 231-100-4

TSCA : Yes

Recommended Storage : Ambient temperatures