Alfa Aesar™ Lead tin oxide dihydrate

Manufacturer: Alfa Aesar™ 2266014

Quantity 25g

 View more versions of this product

Catalog No. AA2266014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12036-31-6
    Linear Formula PbSnO3·2H2O
    Formula Weight 409.94 (373.91 Anhydrous)
    MDL Number MFCD00049760
    UN Number UN3077
    Synonym Lead stannate
    Chemical Name or Material Lead tin oxide dihydrate

    Specifications

    Formula Weight 409.94 (373.91 Anhydrous)
    Health Hazard 3 GHS P Statement
    P273-P201-P308+P313-P501a
    Avoid release to the environment.
    Obtain special instructions before use.
    IF exposed or concerned: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Synonym Lead stannate
    Health Hazard 2 GHS H Statement
    H360-H373-H302-H332
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.
    CAS 12036-31-6
    Color White
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Odor Odorless
    Physical Form Powder
    MDL Number MFCD00049760
    Health Hazard 1 Danger
    TSCA TSCA
    Linear Formula PbSnO3·2H2O
    UN Number UN3077
    Quantity 25g
    Chemical Name or Material Lead tin oxide dihydrate

    GHS H Statement
    H360-H373-H302-H332
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P273-P201-P308+P313-P501a
    Avoid release to the environment.
    Obtain special instructions before use.
    IF exposed or concerned: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit