Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar Lead Tin, solder alloy, 1.6mm (0.06 in.) dia., with rosin core
Manufacturer: Alfa Aesar 4102429
Specifications
Pb:Sn; 50:50 wt% | |
225g | |
MFCD00798549 |
Lead Tin, Solder Alloy | |
1.6mm (0.06 in.) dia., with rosin core | |
Solid |
Safety and Handling
GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
H302+H332-H350-H360Df-H373
TSCA : Yes
Recommended Storage : Ambient temperatures
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title