Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Lead Tin, solder alloy, 3.18mm (0.125 in.) dia.

Manufacturer:  Alfa Aesar™ 4102529

Quantity 225g

 View more versions of this product

Catalog No. AA4102529

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

  • For use with copper, copper alloys, lead, steel, and nickel alloys
  • Recommended for machine and torch soldering
Description & Specifications

Chemical Identifiers

MFCD00798549
Lead Tin, Solder Alloy, 3.18mm (0.125 in.) dia.

Specifications

Pb:Sn; 70:30 wt%
225g
MFCD00798549
Lead Tin, Solder Alloy, 3.18mm (0.125 in.) dia.

Safety and Handling

SDS & Safety Documents

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P273-P201-P308+P313-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit