Alfa Aesar™ Lead Tin, solder alloy, 3.18mm (0.125 in.) dia.

Manufacturer: Alfa Aesar™ 41025TB

Quantity 113g

 View more versions of this product

Catalog No. AA41025TB

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00798549
    Chemical Name or Material Lead Tin, Solder Alloy, 3.18mm (0.125 in.) dia.

    Specifications

    Health Hazard 3 GHS P Statement
    P273-P201-P308+P313-P501a
    Avoid release to the environment.
    Obtain special instructions before use.
    IF exposed or concerned: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    MDL Number MFCD00798549
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H360-H373-H302-H332
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.
    Avg. Mol. Wt. or Mol. Wt. Range Pb:Sn; 70:30 wt%
    Quantity 113g
    Chemical Name or Material Lead Tin, Solder Alloy, 3.18mm (0.125 in.) dia.

    Description

    • For use with copper, copper alloys, lead, steel, and nickel alloys
    • Recommended for machine and torch soldering

    GHS H Statement
    H360-H373-H302-H332
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P273-P201-P308+P313-P501a
    Avoid release to the environment.
    Obtain special instructions before use.
    IF exposed or concerned: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit