Alfa Aesar™ Lead titanium oxide, polymeric precursor, oxide >30 wt%

Used in electronic devices

Manufacturer: Alfa Aesar™ 3975609

Quantity 10g

 View more versions of this product

Catalog No. AA3975609

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12060-00-3
    MDL Number MFCD00011159
    UN Number UN1993
    Synonym Lead titanate
    Chemical Name or Material Lead titanium oxide, Polymeric precursor

    Specifications

    Physical Form Viscous Liquid
    Health Hazard 2 GHS H Statement
    H225-H360-H373-H302-H332-H319
    Highly flammable liquid and vapor.
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Color Brown
    Quantity 10g
    Sensitivity Moisture Sensitive
    CAS 12060-00-3
    UN Number UN1993
    Flash Point 12° (54°F)
    Solubility Information Soluble in alcohols
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    MDL Number MFCD00011159
    Synonym Lead titanate
    Health Hazard 1 Danger
    Concentration Oxide >30 wt%
    Chemical Name or Material Lead titanium oxide, Polymeric precursor

    GHS H Statement
    H225-H360-H373-H302-H332-H319
    Highly flammable liquid and vapor.
    May damage fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful if inhaled.
    Causes serious eye irritation.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit