Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Lead titanium oxide, polymeric precursor, oxide >30 wt%

Used in electronic devices

Manufacturer:  Alfa Aesar™ 3975618

Quantity 50g

 View more versions of this product

Catalog No. AA3975618


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

12060-00-3
UN1993
Lead titanium oxide, Polymeric precursor
MFCD00011159
Lead titanate

Specifications

12060-00-3
MFCD00011159
Lead titanate
Oxide >30 wt%
Brown
12° (54°F)
Viscous Liquid
UN1993
Soluble in alcohols
Moisture Sensitive
50g
Lead titanium oxide, Polymeric precursor

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H360-H373-H302-H332-H319
Highly flammable liquid and vapor.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit