Lead Wire, 0.5mm (0.02 in.) dia., 99.998% (metals basis), Alfa Aesar™, Puratronic™

Form: approx. 2.23g/m

Manufacturer: Alfa Aesar 11902G1

Quantity 1m

 View more versions of this product

Catalog No. AA11902G1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7439-92-1
    Physical Form Wire, 0.5mm (0.02 in.) dia.
    Assay Percent Range 99.998% (Metals basis)
    MDL Number MFCD00134050
    Chemical Name or Material Lead

    Specifications

    Quantity 1m
    Color Gray
    Density 11.34g/cm3 at 20°C
    EINECS Number 231-100-4
    Physical Form Wire, 0.5mm (0.02 in.) dia.
    Assay Percent Range 99.998% (Metals basis)
    MDL Number MFCD00134050
    CAS 7439-92-1
    Odor Odorless
    Health Hazard 1 H360-H373-H400-H410-H302-H332: May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful if inhaled.
    Appearance Gray wire
    Chemical Name or Material Lead

    H360-H373-H400-H410-H302-H332: May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful if inhaled.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit