Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Lead zirconium titanium oxide, polymeric precursor, oxide »30 wt%

Used in piezoelectric devices and electro-optic devices

Manufacturer:  Alfa Aesar™ 3975830

Quantity 250g

 View more versions of this product

Catalog No. AA3975830


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

12626-81-2
MFCD00212270
UN1993
Lead zirconium titanium oxide, Polymeric precursor

Specifications

12626-81-2
MFCD00212270
Soluble in alcohols
Pale yellow
Lead zirconium titanium oxide, Polymeric precursor
Semi-Solid/Viscous Liquid
UN1993
Oxide ≈30 wt%
250g

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H360-H304-H373-H318-H302-H336-H315
Highly flammable liquid and vapor.
May damage fertility or the unborn child.
May be fatal if swallowed and enters airways.
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye damage.
Harmful if swallowed.
May cause drowsiness or dizziness.
Causes skin irritation.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit