Learn More
BiolegendSupplier Diversity Partner LEAF™ Purified anti-human CD336 (NKp44) DFS Item
Manufacturer:  BiolegendSupplier Diversity Partner 325104
LEAF™ Purified anti-human CD336 (NKp44) [P44-8]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC, Block; Size: 500 ug
Catalog No. 50-167-265