Learn More
Impacto™ Leather Full Finger Anti-Impact Carpal Tunnel Glove
Impacto™ Leather Full Finger Anti-Impact Carpal Tunnel Gloves have a unique channel over the carpal tunnel to keep pressure off the median nerve
Manufacturer:  Impacto™ ST501020
 View more versions of this product
Catalog No. 19-400-097
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Provides protection from vibration and impact
  • Visco-elastic polymer (VEP) padding dissipates shock
  • Thin, snug-fit padding is precision-cut to eliminate bunching, and provide protection
  • Fingers are gusseted for comfort
  • Utilizes patented Shock-Tek technology
  • VEP palm padding protects from impact
  • Protected against repetitive strain
Specifications
Specifications
Cowhide Leather
Small
White
CE Certified