Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Impacto™ Full-Finger Anti-Impact Cowhide Leather Carpal Tunnel Gloves
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Description
- Helps prevent the onset of injuries such as carpal tunnel syndrome, repetitive strain, and white finger disease
- Thin, snug fit padding is precision-cut to eliminate bunching and increase comfort
- Full finger design extends protection throughout the entire hand
- VEP padding in the palm minimizes the effects of shock and impact
- Ideal for construction, packaging, material handling, and assembly line work
Specifications
Color | Natural |
Material | Cowhide Leather |
Product Type | Anti-impact Gloves |
Size | Medium |
For Use With (Application) | Prevention of Repetative Strain Injuries (RSI) |
Glove Cuff Style | Elasticated |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction