Learn More
LECT2 Mouse anti-Human, Alexa Fluor 488, Clone: MM0449-4B24, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP211768AF488
Catalog No. NB211768488
Add to cart
Description
Description
LECT2 Monoclonal specifically detects LECT2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
LECT2
MM0449-4B24
Western Blot, Immunohistochemistry
Purified
LECT2
Human LECT2
Protein G purified
Store at 4C in the dark.
Monoclonal
Human
Western Blot, Immunohistochemistry
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
chm2, chm-II, chondromodulin-II, hLECT2, LECT-2, leukocyte cell-derived chemotaxin 2, leukocyte cell-derived chemotaxin-2, MGC126628
Mouse
IgG1
0.1 ml
Primary
3950