Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LECT2 Mouse anti-Human, Clone: MM0449-4B24, Novus Biologicals™
Mouse Monoclonal Antibody
$177.16 - $493.00
Specifications
Antigen | LECT2 |
---|---|
Clone | MM0449-4B24 |
Concentration | LYOPH |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Description
LECT2 Monoclonal specifically detects LECT2 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
LECT2 | |
LYOPH | |
Monoclonal | |
Purified | |
RUO | |
O14960 | |
3950 | |
IgG1 | |
Protein G purified |
MM0449-4B24 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
chm2, chm-II, chondromodulin-II, hLECT2, LECT-2, leukocyte cell-derived chemotaxin 2, leukocyte cell-derived chemotaxin-2, MGC126628 | |
LECT2 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title