Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LECT2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA560541

Catalog No. PIPA560541


Add to cart

Description

Description

Immunogen sequence: NKNAINNGVR ISGRGFCVKM FYIKPIKYKG PIKKGEKLGT LLPLQKVYPG IQSHVHIENC DSSDPT Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.

This gene encodes a secreted, 16 kDa protein that acts as a chemotactic factor to neutrophils and stimulates the growth of chondrocytes and osteoblasts. This protein has high sequence similarity to the chondromodulin repeat regions of the chicken myb-induced myeloid 1 protein. A polymorphism in this gene may be associated with rheumatoid arthritis.
Specifications

Specifications

LECT2
Polyclonal
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O14960
Leukocyte cell-derived chemotaxin 2; hLECT2; LECT2
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
3950
Immunohistochemistry
Unconjugated
LECT2
Liquid
LECT2
Recombit protein corresponding to Human LECT2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Research Use Only