Learn More
Lectin, Wheat Germ Agglutinin from Triticum vulgaris , MP Biomedical
Catalog No. p-7112941
Click to view available options
Quantity:
25 mg
100 mg
Lectin, Wheat Germ Agglutinin from Triticum vulgaris are proteins or glycoproteins of non-immune origin that agglutinate cells and/or precipitate complex carbohydrates.
  • Lectins are very valuable in a wide variety of applications in vitro, including blood grouping and erythrocyte polyagglutination studies, Mitogenic stimulation of lymphocytes, Lymphocyte subpopulation studies, Fractionation of cells and other particles, Histochemical studies of normal and pathological conditions
  • Lectins are capable of binding glycoproteins even in presence of various detergents
  • The agglutination activity of these highly specific carbohydrate-binding molecules is usually inhibited by a simple monosaccharide, but for some lectins, di, tri, and even polysaccharides are required
  • It is purified Triticum vulgrislectin (WGA) from Wheat Germ
  • WGA is not blood group specific but has an affinity for N-acetyl-β-D-glucosaminyl residues and N-acetyl-β-D-glucosamine oligomers
  • WGA contains no protein-bound carbohydrate
TRUSTED_SUSTAINABILITY
Specifications
Color White
Quantity 25 mg
Packaging Glass
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.