Learn More
Lecture Bottle Stand and Carrier
SCI_ED
Keep cylinders safe and secure.
$70.61 - $202.65
Specifications
Material PVC
For Use With (Equipment) Gas cylinder
View More Specs
Products 2
Catalog Number Mfr. No. Dimensions (L x W x H) Holds Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W x H) Holds Product Type Price Quantity & Availability  
S436781
SCI_ED
View Documents
Justrite Manufacturing Co
35316/EMD
8 x 8 x 5 in. (20.3 x 20.32 x 12.7 cm) One round bottom cylinder Lecture Bottle Stand
Each for $70.61
Add to cart
S436782
SCI_ED
View Documents
Justrite Manufacturing Co
35322/EMD
18.5 x 8.6 x 5.75 in. (47 x 21.84 x 14.6 cm) Up to six cylinders Lecture Bottle Carrier
Each for $202.65
Add to cart
Description
Description
Heavy-duty PVC stand and carrier are designed to safely hold lecture bottles.
Carrier
  • Makes it easy to transport up to six cylinders between classrooms
  • Dimensions: 18.5 × 8.6× 5.75 in. (46.3× 21.6× 14.4cm)
Stand
  • Holds the round bottom cylinders upright, keeping your hands free for dispensing and filling
  • Dimensions: 8 × 8 × 5 in. (20 × 20 × 12.5cm)
Specifications
Specifications
PVC
Gas cylinder